Learn More
Invitrogen™ ERCC6L Polyclonal Antibody

Description
The antibody detects endogenous levels of total ERCC6L protein.
Specifications
Specifications
| Antigen | ERCC6L |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Ercc6l |
| Gene Accession No. | Q2NKX8 |
| Gene Alias | ATP-dependent helicase ERCC6-like; BC004701; D330021P09Rik; DNA excision repair protein ERCC-6-like; ERCC excision repair 6 like, spindle assembly checkpoint helicase; ERCC6L; excision repair cross-complementation group 6 like; excision repair cross-complementation group 6-like; excision repair cross-complementing rodent repair deficiency complementation group 6 - like; excision repair cross-complementing rodent repair deficiency complementation group 6 like; excision repair cross-complementing rodent repair deficiency complementation group 6-like; excision repair cross-complementing rodent repair deficiency, complementation group 6-like; excision repair protein ERCC6-like; FLJ20105; LOW QUALITY PROTEIN: DNA excision repair protein ERCC-6-like; MGC131695; PICH; PLK1-interacting checkpoint helicase; RAD26L; RGD1565734; SNF2/RAD54 family protein; tumor antigen BJ-HCC-15 |
| Gene Symbols | Ercc6l |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.