Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERCC6L Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987226
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7226 Supplier Bioss Supplier No. BS6380R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERCC6L is a DNA helicase that acts as an essential component of the spindle assembly checkpoint. ERCC6L contributes to the mitotic checkpoint by recruiting MAD2 to kinetochores and monitoring tension on centromeric chromatin. ERCC6L acts as a tension sensor that associates with catenated DNA which is stretched under tension until it is resolved during anaphase.

Specifications

Antigen ERCC6L
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Ercc6l
Gene Accession No. Q2NKX8
Gene Alias ATP-dependent helicase ERCC6-like; BC004701; BOS_25822; D330021P09Rik; DNA excision repair protein ERCC-6-like; ERCC excision repair 6 like, spindle assembly checkpoint helicase; ERCC6L; excision repair cross-complementation group 6 like; excision repair cross-complementation group 6-like; excision repair cross-complementing rodent repair deficiency complementation group 6 - like; excision repair cross-complementing rodent repair deficiency complementation group 6 like; excision repair cross-complementing rodent repair deficiency complementation group 6-like; excision repair cross-complementing rodent repair deficiency, complementation group 6-like; excision repair protein ERCC6-like; FLJ20105; MGC131695; PICH; PLK1-interacting checkpoint helicase; RAD26L; RGD1565734; SNF2/RAD54 family protein; Tumor antigen BJ-HCC-15
Gene Symbols Ercc6l
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ERCC6L.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54821
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.