Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERCC8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | ERCC8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ERCC8 Polyclonal specifically detects ERCC8 in Human samples. It is validated for Western Blot.Specifications
ERCC8 | |
Polyclonal | |
Rabbit | |
Cancer, DNA Repair, Nucleotide Excision Repair | |
Q13216 | |
1161 | |
IgG | |
44 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
CKN1DNA excision repair protein ERCC-8, Cockayne syndrome 1 (classical), Cockayne syndrome WD repeat protein CSA, CSACockayne syndrome WD-repeat protein CSA, excision repair cross-complementing rodent repair deficiency, complementationgroup 8 | |
ERCC8 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title