Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERCC8 Antibody (OTI2G1), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270412B
Description
ERCC8 Monoclonal specifically detects ERCC8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ERCC8 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
ERCC8 | |
Full length recombinant protein of human ERCC8 (NP_001007235) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI2G1 | |
Western Blot, Immunohistochemistry | |
CKN1DNA excision repair protein ERCC-8, Cockayne syndrome 1 (classical), Cockayne syndrome WD repeat protein CSA, CSACockayne syndrome WD-repeat protein CSA, excision repair cross-complementing rodent repair deficiency, complementationgroup 8 | |
Mouse | |
Affinity Purified | |
Cancer, DNA Repair, Nucleotide Excision Repair | |
1161.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction