Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ERCC8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580596
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ERCC8.
This gene encodes a WD repeat protein, which interacts with Cockayne syndrome type B (CSB) protein and with p44 protein, a subunit of the RNA polymerase II transcription factor IIH. Mutations in this gene have been identified in patients with hereditary disease Cockayne syndrome (CS). CS cells are abnormally sensitive to ultraviolet radiation and are defective in the repair of transcriptionally active genes.
Specifications
ERCC8 | |
Polyclonal | |
Unconjugated | |
ERCC8 | |
2410022P04Rik; 2810431L23Rik; 4631412O06Rik; B130065P18Rik; Ckn1; Cockayne syndrome 1 (classical); Cockayne syndrome 1 homolog; cockayne syndrome WD repeat protein CSA; Cockayne syndrome WD repeat protein CSA homolog; Cockayne syndrome WD-repeat protein CSA; CSA; DNA excision repair protein ERCC-8; ERCC excision repair 8, CSA ubiquitin ligase complex subunit; Ercc8; excision repair cross-complementation group 8; excision repair cross-complementing rodent repair deficiency complementation group 8; excision repair cross-complementing rodent repair deficiency, complementation group 8; excision repaiross-complementing rodent repair deficiency, complementation group 8; UVSS2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1161 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with no preservative | |
Q13216 | |
ERCC8 | |
A synthetic peptide corresponding to the C-terminus of the human ERCC8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction