Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERF/Ets2 Repressor Factor Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982610
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2610 Supplier Bioss Supplier No. BS13096R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Members of the ETS family of transcription factors, such as ERF, regulate cell proliferation and differentiation. They share a highly conserved DNA-binding domain, the ETS domain, that recognizes the sequence GGAA/T (de Castro et al., 1997 [PubMed 9192842]). For further information on ETS transcription factors, see ETS1 (MIM 164720).

Specifications

Antigen ERF/Ets2 Repressor Factor
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ERF
Gene Accession No. P50548
Gene Alias BOS_17751; CRS4; ERF; Est2 repressor factor; ETF1; ETS domain-containing transcription factor ERF; Ets2 repressor factor; Ets2 repressor factor; ets2 repressor factor; eukaryotic translation termination factor 1; fc22h01; PE 2; PE2; PE-2; wu:fc22h01
Gene Symbols ERF
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ERF/Ets2 Repressor Factor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2077
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.