Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584773 Supplier Invitrogen™ Supplier No. PA584773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSSSLFSAVV ARRLGRGSVS DCSDGTSELE EPLGEDPRAR PPGPPDLGAF RGPPLARLPH DPGVF Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

Members of the ETS family of transcription factors, such as ERF, regulate cell proliferation and differentiation. They share a highly conserved DNA-binding domain, the ETS domain, that recognizes the sequence GGAA/T (de Castro et al., 1997 [PubMed 9192842]). For further information on ETS transcription factors, see ETS1 (MIM 164720).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERF
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERF
Gene Accession No. P50548
Gene Alias CRS4; ERF; Est2 repressor factor; ETS domain-containing transcription factor ERF; ETS domain-containing transcription factor ERF; ets2 repressor factor; Ets2 repressor factor; fc22h01; LOW QUALITY PROTEIN: ETS domain-containing transcription factor ERF; PE 2; PE2; PE-2; wu:fc22h01
Gene Symbols ERF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ERF. Recombinant protein control fragment (Product #RP-103876).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2077
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.