Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eRF1 Antibody (OTI2E3), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270692AF750
Description
eRF1 Monoclonal specifically detects eRF1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry.Specifications
eRF1 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ETF1 | |
Full length recombinant protein of human eRF1 (NP_004721) produced in HEK293T cells. | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2a |
Western Blot, Flow Cytometry | |
OTI2E3 | |
Western Blot, Flow Cytometry | |
D5S1995, ERF1eRF1, eukaryotic peptide chain release factor subunit 1, Eukaryotic release factor 1, eukaryotic translation termination factor 1, polypeptide chain release factor 1, Protein Cl1, RF1ERF, sup45 (yeast omnipotent suppressor 45) homolog-like 1, SUP45L1, TB3-1MGC111066 | |
Mouse | |
0.1 mL | |
2107 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction