Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eRF1 Antibody (OTI2E3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200740
Description
eRF1 Monoclonal specifically detects eRF1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry.Specifications
eRF1 | |
Monoclonal | |
0.97 mg/mL | |
Western Blot 1:500, Flow Cytometry 1:100 | |
ETF1 | |
Full length human recombinant protein of human ETF1 (NP_004721) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
2107 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry | |
OTI2E3 | |
Unconjugated | |
D5S1995, ERF1eRF1, eukaryotic peptide chain release factor subunit 1, Eukaryotic release factor 1, eukaryotic translation termination factor 1, polypeptide chain release factor 1, Protein Cl1, RF1ERF, sup45 (yeast omnipotent suppressor 45) homolog-like 1, SUP45L1, TB3-1MGC111066 | |
Mouse | |
48.9 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction