Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eRF1 Antibody (OTI2E3), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270692PCP
Description
eRF1 Monoclonal specifically detects eRF1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.Specifications
| eRF1 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| ETF1 | |
| Full length recombinant protein of human eRF1 (NP_004721) produced in HEK293T cells. | |
| 0.1 mL | |
| 2107.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2E3 | |
| Flow Cytometry | |
| D5S1995, ERF1eRF1, eukaryotic peptide chain release factor subunit 1, Eukaryotic release factor 1, eukaryotic translation termination factor 1, polypeptide chain release factor 1, Protein Cl1, RF1ERF, sup45 (yeast omnipotent suppressor 45) homolog-like 1, SUP45L1, TB3-1MGC111066 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction