Learn More
Invitrogen™ ERG Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
$459.00 - $1159.00
Specifications
Antigen | ERG Biosimilar |
---|---|
Concentration | 1 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
Classification | Recombinant Monoclonal |
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Jackson Foundation patent anti-ERG.
v-ets erythroblastosis virus E26 oncogene like isoform 2 (ERG) is a transcriptional regulator. It may participate in transcriptional regulation through the recruitment of SETDB1 histone methyltransferase and subsequent modification of local chromatin structure.Specifications
ERG Biosimilar | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Lyophilized | |
IgG1 | |
Human | |
Primary | |
Protein A |
1 mg/mL | |
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
Unconjugated | |
Human | |
RUO | |
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.