Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERG Mouse anti-Human, Clone: OTI8A9, liquid, TrueMAB™
SDP

Catalog No. 501677250
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7250 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7250 Supplier OriGene Supplier No. TA600177
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Transcriptional regulator. May participate in transcriptional regulation through the recruitment of SETDB1 histone methyltransferase and subsequent modification of local chromatin structure.

Specifications

Antigen ERG
Applications ELISA
Classification Monoclonal
Clone OTI8A9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 10mg/ml BSA, 50% glycerol and 0.05% sodium azide
Gene ERG
Gene Accession No. P11308
Gene Alias erg-3, p55
Gene Symbols ERG
Host Species Mouse
Immunogen Full length human recombit protein of human ERG produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2078
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.