Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549681
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549681 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549681 Supplier Invitrogen™ Supplier No. PA549681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous level of total ERG protein.

v-ets erythroblastosis virus E26 oncogene like isoform 2 (ERG) is a transcriptional regulator. It may participate in transcriptional regulation through the recruitment of SETDB1 histone methyltransferase and subsequent modification of local chromatin structure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERG
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Erg
Gene Accession No. P11308, P81270
Gene Alias avian erythroblastosis virus E-26 (v-ets) oncogene related; D030036I24Rik; ERG; ERG, ETS transcription factor; ERG/EWS fusion gene; ERG/FUS fusion gene; ERG/TMPSSR2 fusion gene; ERG1; Erg-3; erythroblast transformation-specific transcription factor ERG variant 10; ETS transcription factor ERG; ets-related; H Kv11.3; HGNC:3446; KCNH2; Oncogene ERG; OTTHUMP00000115970; OTTHUMP00000208036; p55; TMPRSS2/ERG fusion; transcriptional regulator ERG; transcriptional regulator ERG (transforming protein ERG); transcriptional regulator ERG; LOW QUALITY PROTEIN: transcriptional regulator ERG; Transforming protein ERG; v-ets avian erythroblastosis virus E26 oncogene; v-ets avian erythroblastosis virus E26 oncogene homolog; v-ets avian erythroblastosis virus E26 oncogene related; v-ets erythroblastosis virus E26 oncogene homolog; v-ets erythroblastosis virus E26 oncogene like
Gene Symbols Erg
Host Species Rabbit
Immunogen Peptide sequence around aa.84~88(G-S-R-N-S) derived from Human ERG.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13876, 170909, 2078
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.