Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERG Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206949
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6949 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6949 Supplier Fabgennix Inc Supplier No. ERG101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERG Polyclonal antibody specifically detects ERG in Human, Mouse, Rat samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen ERG
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Erg
Gene Accession No. P11308, P81270
Gene Alias avian erythroblastosis virus E-26 (v-ets) oncogene related; D030036I24Rik; ERG; ERG, ETS transcription factor; Erg-3; erythroblast transformation-specific transcription factor ERG variant 10; ETS transcription factor ERG; ets-related; H Kv11.3; HGNC:3446; OTTHUMP00000115970; OTTHUMP00000208036; p55; TMPRSS2/ERG fusion; transcriptional regulator ERG; transcriptional regulator ERG (transforming protein ERG); transcriptional regulator ERG; LOW QUALITY PROTEIN: transcriptional regulator ERG; Transforming protein ERG; v-ets avian erythroblastosis virus E26 oncogene; v-ets avian erythroblastosis virus E26 oncogene homolog; v-ets avian erythroblastosis virus E26 oncogene related; v-ets erythroblastosis virus E26 oncogene homolog; v-ets erythroblastosis virus E26 oncogene like
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 400-450 on human Transcriptional regulator ERG protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13876, 170909, 2078
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.