Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERGIC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553874
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553874 Supplier Invitrogen™ Supplier No. PA553874
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGNFHVSTHS ATAQPQNPDM THVIHKLSFG DTLQVQNIHG AFNALGGADR LTSNPLASHD YIL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

The function of ERGIC1 remains largely unknown. ERGIC1 was highly expressed in most primary prostate tumors. It is an intriguing potential drug target especially for the ERG oncogene expressing tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERGIC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERGIC1
Gene Accession No. Q969X5
Gene Alias 1200007D18Rik; endoplasmic reticulum-golgi intermediate compartment (ERGIC) 1; endoplasmic reticulum-golgi intermediate compartment 1; endoplasmic reticulum-golgi intermediate compartment 32 kDa protein; endoplasmic reticulum-Golgi intermediate compartment protein 1; ERGIC1; ERGIC32; ERGIC-32; ER-Golgi intermediate compartment 32 kDa protein; HT034; KIAA1181; maa-136; mKIAA1181 protein-like; NET24
Gene Symbols ERGIC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ERGIC1. Recombinant protein control fragment (Product #RP-92411).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57222
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.