Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERGIC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100505
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100505 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100505 Supplier Invitrogen™ Supplier No. PA5100505
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ERGIC2.

ERGIC2, or PTX1, is a ubiquitously expressed nuclear protein that is downregulated in prostate carcinoma (Kwok et al., 2001).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERGIC2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ERGIC2
Gene Accession No. Q96RQ1
Gene Alias 1200009B18Rik; 4930572C01Rik; AA408438; cd002; CD14 protein; CDA14; Endoplasmic reticulum-Golgi intermediate compartment protein 2; ERGIC and golgi 2; ERGIC2; ERV41; PTX1; PTX1 protein; RGD1310606
Gene Symbols ERGIC2
Host Species Rabbit
Immunogen A synthesized peptide derived from human ERGIC2(Accession Q96RQ1), corresponding to amino acid residues F338-H377.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51290
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.