Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERGIC2 Polyclonal Antibody
ERGIC2 Polyclonal Antibody
Supplier: Thermo Scientific PA568643
Description
ERGIC2 Polyclonal Antibody for Western Blot

Specifications
ERGIC2 | |
Polyclonal | |
Unconjugated | |
ERGIC2 | |
1200009B18Rik; 4930572C01Rik; AA408438; cd002; CD14 protein; CDA14; Endoplasmic reticulum-Golgi intermediate compartment protein 2; ERGIC and golgi 2; ERGIC2; ERV41; PTX1; PTX1 protein; RGD1310606 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
67456 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9CR89 | |
ERGIC2 | |
synthetic peptide directed towards the N-terminal region of mouse ERGIC2. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction