Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERGIC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553289
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553289 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553289 Supplier Invitrogen™ Supplier No. PA553289
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CNTCEDVREA YRRRGWAFKN PDTIEQCRRE GFSQKMQEQK NEGCQVYGFL EVNKVAGNFH FAPGKSFQQS HVH Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

ERGIC3 is possible role in transport between endoplasmic reticulum and Golgi.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERGIC3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ERGIC3
Gene Accession No. Q9Y282
Gene Alias 2310015B14Rik; AV318804; C20orf47; CGI-54; D2Ucla1; dJ477O4.2; Endoplasmic reticulum-Golgi intermediate compartment protein 3; endoplasmic reticulum-localized protein ERp43; ERGIC and golgi 3; ERGIC3; ERV46; NY-BR-84; PRO0989; Sdbcag84; serologically defined breast cancer antigen 84; serologically defined breast cancer antigen NY-BR-84; Serologically defined breast cancer antigen NY-BR-84 homolog
Gene Symbols ERGIC3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ERGIC3. Recombinant protein control fragment (Product #RP-90949).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51614
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.