Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NoTrax™ Ergo Trax™ Grande™

Catalog No. p-7214663
Click to view available options
Dimensions (L x W):
2 x 3 ft.
3 x 5 ft.
3 x 12 ft.

Ergo Trax™ Grande™ a thicker, heavier and stronger anti-fatigue floor mat

  • Thicker, heavier, stronger anti-fatigue floor mat
  • For industrial applications and multi-shift operations in dry work areas
  • Thickness: 1 in.
  • Maximizes worker productivity and comfort
  • Durable vinyl diamond-plate top surface design provides non-directional traction
  • Easy to clean
  • Top surface combined with a Niru™ rubber sponge base utilizing NoTrax™ exclusive UniFusion™ technology
    • Eliminates possibility of de-lamination
  • Sloped beveled edges reduce tripping hazards and allows easy access on and off mat

Specifications

Type Anti-fatigue Floor Mat
Dimensions (L x W) 2 x 3 ft.
Length (English) 2 ft.
Material Heavy duty vinyl top surface combined with Niru™ rubber base
Color Black, Yellow
Width (English) 3 ft.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.