Learn More
Ergodyne™ Arsenal™ SCBA Mask Bag
Catalog No. 191300571 Shop All Ergodyne Products
Click to view available options
Material:
Fleece-lined, Nylon, Double-coated Polyurethane
Unlined, Nylon, Double-coated Polyurethane
Catalog No. 19-130-0571
Supplier: Ergodyne™ 13080
Stores and protects SCBA masks.
  • Fits most masks
  • Features drawstring opening and barrel closure
  • Double-coated 1000D nylon repels water and resists abrasions, tears, and punctures
  • Spring-loaded steel clasp attaches to web gear or belt
  • Reflective stripe
  • Available unlined or with brushed poly-fleece lining
Specifications
Product Type SCBA Mask Bag
Color Red
Material Unlined, Nylon, Double-coated Polyurethane
Model 13080
Dimensions (L x W x H) 8.5 x 8.5 x 12 in. (21 x 21 x 31 cm)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.