Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ergodyne™ Chill-Its™ 6605 High-Performance Headband
Shop All Ergodyne Products
Click to view available options
Color:
Navy Western
Stars and Stripes
Description
Technical fabric moves moisture away from body for fast evaporation.
- Made of Hi Cool™ Moisture Management performance fabric
- Elasticized back for a great fit
- Machine washable
Specifications
Specifications
Material | Fabric |
Color | Stars and Stripes |
For Use With (Application) | Construction, trades, maintenance, landscaping/grounds, assembly/fabrication, material handling, freight/baggage, warehousing/distribution, Iron/Steel fabrication, jungle love |
Includes | Elasticized Back |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction