Learn More
Description
- Provide warmth to covered area
- Soothe painful strains
- Support without muscle restriction
- Conform to the body's natural shape
- Made of neoprene four way stretch fabric
- Single layer
- Ambidextrous
- Color: black
Specifications
Specifications
| Color | Black |
| Closure Type | Adjustable cinch strap |
| Product Type | Elbow Sleeve |
| Design | Contoured |
| Size | Small |
| For Use With (Application) | Proper lifting protection, support without muscle restriction, helps soothe painful strains |
| Hand | Ambidextrous |
| Material | Neoprene |
| Model | 650 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.