Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 650 Neoprene Elbow Sleeve

Catalog No. 19037842E Shop All Ergodyne Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
For Use With (Application):
Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis
Proper lifting protection, support without muscle restriction, helps soothe painful strains
Material:
Neoprene
Design:
Contoured
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size For Use With (Application) Material Design
19-037-842E 2X-Large Proper lifting protection, support without muscle restriction, helps soothe painful strains Neoprene Contoured
19-037-842B Medium Proper lifting protection, support without muscle restriction, helps soothe painful strains Neoprene Contoured
19-037-842A Small Proper lifting protection, support without muscle restriction, helps soothe painful strains Neoprene Contoured
19-037-842C Large Proper lifting protection, support without muscle restriction, helps soothe painful strains Neoprene Contoured
19-037-843A Small Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis Neoprene Contoured
19-037-843B Medium Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis Neoprene Contoured
19-037-843C Large Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis Neoprene Contoured
19-037-842D X-Large Proper lifting protection, support without muscle restriction, helps soothe painful strains Neoprene Contoured
19-037-843E 2X-Large Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis Neoprene Contoured
19-037-843D X-Large Proper lifting protection, support without muscle restriction, helps soothe inflamed tissue associated with painful strains and tendinitis Neoprene Contoured
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 19-037-842E Supplier Ergodyne™ Supplier No. 16576

Proper lifting protection

  • Provide warmth to covered area
  • Soothe painful strains
  • Support without muscle restriction
  • Conform to the body's natural shape
  • Made of neoprene four way stretch fabric
  • Single layer
  • Ambidextrous
  • Color: black

Specifications

Color Black
Closure Type Adjustable cinch strap
Product Type Elbow Sleeve
Design Contoured
Size 2X-Large
For Use With (Application) Proper lifting protection, support without muscle restriction, helps soothe painful strains
Hand Ambidextrous
Material Neoprene
Model 650
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.