Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 600/615/620 Knee Sleeves

Catalog No. 19037835C Shop All Ergodyne Products
Click to view available options
Design:
Contoured
Contoured; Open patella/anterior pad
Neoprene four-way stretch Fabric, closed cell structure
Single layer, Neoprene four-way stretch Fabric
Size:
Small
Medium
Large
X-Large
2X-Large
Model:
600
615
620

Specifications

Color Black
Product Type Proflex™ Knee Sleeve
Design Single layer, Neoprene four-way stretch Fabric
Size Large
For Use With (Application) Provide glove-like fit and comfortable wear and provides uniform support and warmth to affected area
Material Neoprene
Model 600
Size (English) 15 to 16 in.
Size (Metric) 38 to 41 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.