Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ 819WP Thermal Waterproof Gloves

Catalog No. 19147503 Shop All Ergodyne Products
Click to view available options
Size:
Small
Medium
Large
X-Large

Gloves feature a breathable waterproof and windproof Hipora liner, as well as 40 grams of 3M™ Thinsulate™ insulation. PVC coating on the palm, fingers, and saddle enhances grip capabilites in damp conditions.

  • Breathable waterproof and windproof Hipora insert liner
  • 3M™ Thinsulate™ thermal insulation for cold weather applications
  • PVC coating on the palm, fingers, and saddle enhances grip and durability
  • EVA knuckle pad with reflective dots for protection against bumps and bruises
  • Gauntlet cuff with pull on tab for easy donning and doffing, especially in wet conditions
  • Terrycloth brow wipe on the back of the thumb

Specifications

Back Material EVA Padding
Coated Area Palm, Fingers, Saddle
Coating Material PVC
Color Black
Hand Specific
Material Nylon, Hipora
Size Medium
For Use With (Application) Cold Storage, Construction, Fishing, Ice Sculpting, Marine Research, and Winter Work
Glove Cuff Style Gauntlet
Liner Material 3M™ Thinsulate™ Insulation (40 g)
Series ProFlex™ 819WP
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.