Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ ProFlex™ Full-Finger Handler Gloves

Catalog No. 19147560 Shop All Ergodyne Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves feature textured silicone on the palm and fingers for enhanced grip capabilities in wet or dry applications.

  • Breathable, 3-layer stretch spandex back
  • Textured silicone palm and fingers
  • Neoprene knuckle pad
  • Low-profile closure with woven elastic cuff
  • Pull tab for easy on and off
  • Terry thumb brow wipe

Specifications

Coating Material Silicone
Color Black
Exterior Finish Partially Textured
Hand Specific
Material Spandex, Neoprene
Product Type Mechanic's Gloves
Size 2X-Large
Finger Style Full
For Use With (Application) Assembly, Distribution, Material Handling, Packaging, and Warehousing
Glove Cuff Style Elasticated
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.