Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids 3400 Glove Clip Holder - Dual Clips

Catalog No. p-3081693 Shop All Ergodyne Products
Click to view available options
Color:
Black
Blue
Orange
This item is not returnable. View return policy
This item is not returnable. View return policy

The strong gripping jaws of Squids Glove Holders provide a secure way to hold gloves, hats, keys, towels, and other large forms of PPE. Dual clip model is ideal for connecting to clothing, belt loops, tool belts, and D-rings.

  • Conveniently holds face masks, gloves, mittens, keys, PPE, towels and more so that they are always in reach
  • Dual clip model is ideal for connecting to clothing, belt loops, tool belts, and D-rings
  • Acetal-copolymer and thermoplastic polyurethane are non-conductive and temperature resistant
  • Patented breakaway design safely separates to allow for individual clips to be used independently
  • Prevents dropped and lost objects, as well as reducing possible injuries caused by bending over to retrieve fallen objects
  • Average breaking strength is 26 lb. (11.8 kg)

Specifications

Color Black
Material Acetal Copolymer, Thermoplastic Polyurethane
Product Type Glove Clip
For Use With (Application) Hanging/Storing PPE
Model Dual Clip
Series 3400
Length (English) 6.5 in.
Length (Metric) 16.5 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.