Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergosterol, 96%, may cont. up to ca 6% water

Catalog No. p-7027244
Click to view available options
Quantity:
5 g
25 g

Store cold | CAS: 57-87-4 | C28H44O | 396.659 g/mol

Ergosterol may be used to study the function of anti-fungal drugs such as Amphotericin B and its analogues and to study the ergosterol biosynthesis pathway within various fungi. It acts as a useful indicator of living fungal biomass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ergosterol may be used to study the function of anti-fungal drugs such as Amphotericin B and its analogues and to study the ergosterol biosynthesis pathway within various fungi. It acts as a useful indicator of living fungal biomass.

Solubility
Soluble in water (<1mg/mL at 25°C), chloroform (50 mg/ml), DMSO (<1mg/mL at 25°C), ethanol (<1mg/mL at 25°C), and ether (14.29 mg/ml).

Notes
Light sensitive. Store away from air, light and oxidizing agents, bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under dark. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-87-4
Molecular Formula C28H44O
MDL Number MFCD00003623
InChI Key DNVPQKQSNYMLRS-APGDWVJJSA-N
PubChem CID 444679
ChEBI CHEBI:16933

Specifications

Melting Point 154°C to 160°C
Assay Percent Range 0.96
Quantity 5 g
UN Number UN2811
Merck Index 14,3659
Synonym ergosterol, provitamin d2, unii-z30ray509f, ergosterin, ergosta-5,7,22e-trien-3beta-ol, 5,7,22-ergostatrien-3beta-ol, 3beta-hydroxy-5,7,22-ergostatriene, 22e,24s-24-methylcholesta-5,7,22-trien-3beta-ol, 22e-ergosta-5,7,22-trien-3beta-ol, ergosta-5,7,22-trien-3-ol, 3b,22e
Solubility Information Soluble in water (<1mg/ml at 25°C),chloroform (50mg/ml),DMSO (<1mg/ml at 25°C),ethanol (<1mg/ml at 25°C),and ether (14.29mg/ml).
SMILES CC(C)C(C)C=CC(C)C1CCC2C1(CCC3C2=CC=C4C3(CCC(C4)O)C)C
IUPAC Name (3S,9S,10R,13R,14R,17R)-17-[(E,2R,5R)-5,6-dimethylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,9,11,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-3-ol
Molecular Weight (g/mol) 396.659
Formula Weight 396.65 (Anhydrous)
Percent Purity 96%
Assay (dry wt.), cont. up to ≈6% water
Chemical Name or Material Ergosterol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H300
Precautionary Statement GHS P Statement
P264-P270-P301+P310-P321-P330-P405-P501a
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 200-352-7
TSCA No
storageNote1 Light sensitive
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.