Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ERH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551723
Description
Immunogen sequence: LVQPTKRPEG RTYADYESVN ECMEGVCKMY EEHLKRMNPN SPSITYDISQ LFDFIDDLAD LSCLVYRADT QTYQPYNKDW IKEKIYVLLR RQAQQAGK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The Enhancer of Rudimentary Homolog (ERH) is a highly conserved protein with significant roles in the regulation of gene expression and cell cycle control. Structurally, ERH forms homodimers, characterized by a crystal structure resolving at 2.0 A, which reveals its ability to bind specific protein regions via a hydrophobic groove. Functionally, ERH interacts with various cellular components, notably integrating into the Microprocessor complex where it facilitates microRNA maturation by binding with the N-terminus of DGCR8 at a 2:2 stoichiometry. This interaction underscores ERH's role in gene silencing, contributing to the biogenesis of small interfering RNAs (siRNAs). Additionally, ERH is implicated in transcriptional regulation and has been associated with DNA replication, suggesting its involvement in broader genomic functions. Despite its diverse roles, ERH operates in a plug-and-play manner, allowing its interaction with different cellular factors to fulfill pleiotropic functions across metazoans.
Specifications
| ERH | |
| Polyclonal | |
| Unconjugated | |
| ERH | |
| DROER; Enhancer of rudimentary homolog; enhancer of rudimentary homolog (Drosophila); Erh; Mer; Prei1; preimplantation protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 13877, 2079, 681415 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P84089, P84090 | |
| ERH | |
| Recombinant protein corresponding to Human ERH. Recombinant protein control fragment (Product #RP-89033). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction