Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517141
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517141 Supplier Thermo Scientific Supplier No. PA517141
Add to Cart
Add to Cart

ERI1 Polyclonal Antibody

ERI1 Polyclonal Antibody for Western Blot, IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen ERI1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene ERI1
Gene Accession No. Q5FVR4, Q7TMF2
Gene Alias 3 ' exoribonuclease; 3' exoribonuclease; 3110010F15Rik; 3'-5' exonuclease ERI1; 3'-5' exoribonuclease 1; 3'EXO; 3'HEXO; enhanced RNAi protein 1; enhanced RNAi three prime mRNA exonuclease homolog 1; ERI1; eri-1; Eri-1 homolog; exoribonuclease 1; HEXO; histone mRNA 3' end-specific exonuclease; histone mRNA 3'-end-specific exoribonuclease; Histone mRNA 3'-exonuclease 1; protei; protein 3'hExo; RGD1308378; Thex1; three prime histone mRNA exonuclease 1; three prime mRNA exonuclease 1
Gene Symbols ERI1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the sequence of mouse THEX1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361159, 67276
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.