Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | ERI1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-9566
![]() ![]() |
Proteintech Group Inc
145921AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
RNA exonuclease that binds to the 3′-end of histone mRNAs and degrades them, suggesting that it plays an essential role in histone mRNA decay after replication. A 2′ and 3′-hydroxyl groups at the last nucleotide of the histone 3′-end is required for efficient degradation of RNA substrates. Also able to degrade the 3′-overhangs of short interfering RNAs (siRNAs) in vitro, suggesting a possible role as regulator of RNA interference (RNAi). Requires for binding the 5′-ACCCA-3′ sequence present in stem-loop structure. Able to bind other mRNAs. Required for 5.8S rRNA 3′-end processing. Also binds to 5.8s ribosomal RNA. Binds with high affinity to the stem-loop structure of replication-dependent histone pre-mRNAs.Specifications
ERI1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q5FVR4, Q7TMF2, Q8IV48 | |
361159, 67276, 90459 | |
ERI1 Fusion Protein Ag6125 | |
Primary | |
-20°C | |
ERI1 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
3 5 exonuclease ERI1, 3 5 exoribonuclease 1, 3EXO, 3′HEXO, Eri 1 homolog, ERI1, exoribonuclease 1, HEXO, Histone mRNA 3 exonuclease 1, Protein 3hExo, THEX1 | |
Eri1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title