Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eriochrome Black T/Sodium Chloride Indicator, Reagents
Click to view available options
Quantity:
100 mL
Specifications
Specifications
Chemical Name or Material | Eriochrome Black T/Sodium Chloride Indicator |
CAS | 7647-14-5, 1787-61-7 |
Boiling Point | 1461°C |
Synonym | Water hardness indicator |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 2.17 |
Color | Gray |
Quantity | 100 mL |
Physical Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction