Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Eriochrome∣r Blue Black B

Catalog No. aaj6339622
Click to view available options
Quantity:
100 g
500 g

Catalog No. AAJ6339622

Supplier: Thermo Scientific Chemicals J6339622

Only null left
Add to Cart

CAS: 3564-14-5 | C20H13N2NaO5S | 416.38 g/mol

Eriochrome is to do titration indicator. Dubbed in aqueous solution or alcohol-soluble when used for the determination of calcium, magnesium, manganese, zinc and other metal ions, measured water hardness.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eriochrome is to do titration indicator. Dubbed in aqueous solution or alcohol-soluble when used for the determination of calcium, magnesium, manganese, zinc and other metal ions, measured water hardness.

Solubility
Soluble in water(20 g/L (20 C).

Notes
Eriochrome is a registered trademark of Ciba Specialty Chemicals Corp.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3564-14-5
Molecular Formula C20H13N2NaO5S
Molecular Weight (g/mol) 416.38
MDL Number MFCD00003934
InChI Key YRFCCVCSEVISHI-GXTSIBQPSA-M
Synonym C.I. 14640; Mordant Black 3
PubChem CID 23722702
SMILES [Na+].OC1=C2C=CC=CC2=CC=C1N\N=C1/C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O

Specifications

Solubility Information Soluble in water(20g/L (20 C).
IUPAC Name sodium;4-[(1-hydroxynaphthalen-2-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate
Formula Weight 416.39
Color Black to Brown
Quantity 100 g
Physical Form Powder
Chemical Name or Material Eriochrome™ Blue Black B
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 222-639-6
RTECSNumber QK2195000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.