Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eriochrome Red B, TCI America™
SDP

Catalog No. E019925G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 3618-63-1
Molecular Formula C20H15N4NaO5S
Molecular Weight (g/mol) 446.413
MDL Number MFCD00004080
InChI Key FPSFKBGHBCHTOE-UHFFFAOYSA-M
Synonym Mordant Red 7
PubChem CID 101586668
IUPAC Name sodium;1-[(3-methyl-5-oxo-1-phenyl-4H-pyrazol-4-yl)diazenyl]-4-sulfonaphthalen-2-olate
SMILES CC1=NN(C(=O)C1N=NC2=C(C=C(C3=CC=CC=C32)S(=O)(=O)O)[O-])C4=CC=CC=C4.[Na+]

Specifications

Formula Weight 446.41
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Eriochrome Red B
EINECSNumber (5)-2062
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.