Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERK1/2 Antibody (B20-U), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP1301280.1ML
Description
ERK1/2 Monoclonal specifically detects ERK1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
ERK1/2 | |
Monoclonal | |
4.0mg/mL | |
Western Blot 1:5000, ELISA 1:20000-1:100000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, p44mapk, PRKM3EC 2.7.11.24 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Major clone of IgGs representing both, Erk1 (Gene ID: 5595, UniProt: P27361) and Erk2 (Gene ID: 5594, UniProt: P28482), obtained from immunoaffinity purified immunoglobulins corresponding to immunogenic peptide. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
B20-U | |
Unconjugated | |
20mM Tris/HCl (pH 8.0) and 10 mg/ml BSA with 0.05% Sodium Azide | |
MAPK3 | |
Peptide derived from C-terminal sequence of human Erk 2. | |
0.1 mL | |
Angiogenesis, Breast Cancer, Cancer, Cell Cycle and Replication, Diabetes Research, Hypoxia, MAP Kinase Signaling, Signal Transduction | |
5595 | |
Human, Mouse, Rat, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction