Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK1/2, p Thr202, p Tyr204, p Thr185, p Tyr187 Antibody (SC58-01), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP267555

Catalog No. NBP267555


Only null left
Add to Cart

Description

Description

ERK1/2 Monoclonal specifically detects ERK1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

ERK1/2 (p Thr202, p Tyr204, p Thr185, p Tyr187)
Monoclonal
1 mg/mL
Western Blot 1:500-1:2000, Immunohistochemistry 1:400, Immunoprecipitation, Immunohistochemistry-Paraffin 1:1000
EC 2.7.11, ERK-1, ERK1p44-MAPK, ERT2, Extracellular signal-regulated kinase 1, extracellular signal-related kinase 1, HS44KDAP, HUMKER1A, Insulin-stimulated MAP2 kinase, MAP kinase 1, MAP kinase 3, MAP kinase isoform p44, MAPK 1, MAPK 3, MGC20180, Microtubule-associated protein 2 kinase, Mitogen-activated protein kinase 1, mitogen-activated protein kinase 3, p44erk1, p44-ERK1, p44mapk, PRKM3EC 2.7.11.24
Rabbit
Protein A purified
Alzheimers Research, Angiogenesis, Apoptosis, Breast Cancer, Cancer, Caspases, Cell Cycle and Replication, Diabetes Research, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Neurodegeneration, Phospho Specific, Protein Kinase, Signal Transduction
5595
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Immunoprecipitation, Western Blot
SC58-01
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
MAPK3
Synthetic peptide within Human aa 166-215/ 379. (SwissProt: P27361 Human; SwissProt: P28482 Human; SwissProt: P63085 Mouse; SwissProt: Q63844 Mouse; SwissProt: P21708 Rat; SwissProt: P63086 Rat)
100 μL
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.