Learn More
ERK1/2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 510681AP150UL

Description
This antibody can recognize both ERK1 and ERK2 with the molecular mass of 38-43 kDa.
MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. The p44/42 MAPK (Erk1/2) are members of mitogen-activated kinases (MAPKs) of serine/threonine protein kinases. Phospho ERK1/2 can be activated by a range of extracellular stimuli, such as mitogen, growth factors, neurotransmitters, chemokines, and cytokines, through receptor tyrosine kinases (RTK), G protein-coupled receptors (GPCRs), or protein kinase C (PKC). Upstream MEK1 and MEK2 activate p44 and p42 (phospho ERK1/2) through phosphorylation of activation loop residues Thr202/Tyr204 and Thr185/Tyr187, respectively. p44/42 (ERK1/2) are negatively regulated by a family of dual-specificity (Thr/Tyr) MAPK phosphatases, known as DUSPs or MKPs, along with MEK inhibitors, such as U0126 and PD98059.Specifications
ERK1/2 | |
Polyclonal | |
Unconjugated | |
MAPK1 | |
ERK, ERK 2, ERK1/2, ERK2, ERT1, MAP kinase 1, MAP kinase 2, MAP kinase isoform p42, MAPK 1, MAPK 2, MAPK1, MAPK2, p40, p41, p41mapk, p42 MAPK, P42MAPK, PRKM1, PRKM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116590, 26413, 26417, 50689, 5594, 5595 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P21708, P27361, P28482, P63085, P63086, Q63844 | |
Mapk1, Mapk3 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.