Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERK1/ERK2, p Thr202 Antibody (SR1701), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32158850UL
Description
ERK1/ERK2 Monoclonal antibody specifically detects ERK1/ERK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
ERK1/ERK2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
A synthesized peptide derived from human phospho-ERK1 [Thr202]/ERK2 [Thr185] (Uniprot #: P27361) | |
50 μg | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
SR1701 | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:40 | |
Rabbit | |
Affinity purified | |
RUO | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction