Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERK1/ERK2, p Tyr204, p Tyr187, p Thr202, p Thr185 Antibody (269434), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB1018S
Description
ERK1/ERK2 Monoclonal antibody specifically detects ERK1/ERK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
ERK1/ERK2 | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Phosphopeptide containing ERK1 T202/Y204 site | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
269434 | |
Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction