Learn More
Promega ERK1 Kinase Enzyme System, 10microg
Catalog No. PRV1951 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV1951 10 μg
1 options
Catalog No. PRV1951 Supplier Promega Supplier No. V1951
Recombinant full-length, tag-free human ERK1 was expressed by baculovirus in Sf9 insect cells and activated by active MEK1 in vitro. ERK1 is a protein serine/threonine kinase that is a member of the extracellular signal-regulated kinases (ERKs).
  • Easily Screen and Profile ERK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage 2°C to 10°C