Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ERK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674625
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-625 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-625 Supplier Invitrogen™ Supplier No. PA19041
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

MAPK3 (ERK1) is a serine/threonine kinase that is an important activator of p90, RSK, MSK, ELK1, and Stat3, and is involved in the MAPK pathway. ERK1 is widely expressed and involved in the regulation of meiosis, mitosis, and post-mitotic functions in differentiated cells. Many different stimuli, including growth factors, cytokines, virus infection, ligands for heterotrimeric guanine nucleotide-binding protein (G protein)-coupled receptors and transforming agents, activate the ERK1 and ERK2 pathways. Functionally, ERK1 is involved with a signaling cascade that regulates various cellular processes such as proliferation, differentiation, and cell cycle progression in response to a variety of extracellular signals. ERK1 is activated by upstream kinases, resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Alternatively spliced transcript variants encoding different protein isoforms of ERK1 have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERK1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MAPK3
Gene Accession No. P27361
Gene Alias ERK; Erk1; ERK-1; Ert2; Esrk1; Extracellular signal-regulated kinase 1; extracellular signal-related kinase 1; HS44KDAP; HUMKER1A; Insulin-stimulated MAP2 kinase; MAP kinase 1; MAP kinase 3; MAP kinase isoform p44; MAP kinase2; MAPK 1; MAPK 3; MAPK1; MAPK2; MAPK3; Microtubule-associated protein 2 kinase; mitogen activated protein kinase 3; mitogen-activated 3; mitogen-activated protein kinase 1; mitogen-activated protein kinase 3; MNK1; Mtap2k; p44; p44 MAP kinase; P44ERK1; p44-ERK1; P44MAPK; p44-MAPK; pp42/MAP kinase; PRKM3
Gene Symbols MAPK3
Host Species Goat
Immunogen Synthetic peptide corresponding to the N terminus amino acids GGEPRRTEGVGP (aa 11-23).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5595
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.