Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK1 Polyclonal Antibody, Huabio

Catalog No. 502544759
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4759 Supplier HUABIO RESEARCH INC Supplier No. ER131011
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ERK1 Polyclonal antibody specifically detects ERK1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ERK1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MAPK3
Gene Accession No. P21708, P27361, Q63844
Gene Alias ERK; Erk1; ERK-1; Ert2; Esrk1; Extracellular signal-regulated kinase 1; extracellular signal-related kinase 1; HS44KDAP; HUMKER1A; Insulin-stimulated MAP2 kinase; MAP kinase 1; MAP kinase 3; MAP kinase isoform p44; MAP kinase2; MAPK 1; MAPK 3; MAPK1; MAPK2; MAPK3; Microtubule-associated protein 2 kinase; mitogen activated protein kinase 3; mitogen-activated 3; mitogen-activated protein kinase 1; mitogen-activated protein kinase 3; MNK1; Mtap2k; p44; p44 MAP kinase; P44ERK1; p44-ERK1; P44MAPK; p44-MAPK; pp42/MAP kinase; PRKM3
Host Species Rabbit
Immunogen Synthetic peptide within human ERK1 aa 306-360.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26417, 50689, 5595
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.