Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERK2 Antibody (1A4), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP174025JF646
Description
ERK2 Monoclonal specifically detects ERK2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ERK2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAPK1 | |
Recombinant human MAPK1 (1-360) purified from E.coli | |
0.1 mL | |
Primary | |
Anti-human MAPK1 mAb, clone 1A4, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human MAPK1 protein. | |
Store at 4°C in the dark. | |
IgG2b κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1A4 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2 | |
Mouse | |
Protein G purified | |
Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Protein Kinase, Signal Transduction, Tumor Suppressors | |
5594 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction