Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERK2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119102JF646
Description
ERK2 Polyclonal antibody specifically detects ERK2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ERK2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2 | |
The immunogen recognized by this antibody maps to a region between residue 310 and 360 of human extracellular signal-regulated kinase-2 (mitogen-activated protein kinase 1) using the numbering given in entry NP_002736.3 (GeneID 5594). | |
0.1 mL | |
Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Tumor Suppressors | |
5594 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction