Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ERK2 Antibody (PCRP-MAPK1-1D1), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314157JF646
This item is not returnable.
View return policy
Description
ERK2 Monoclonal antibody specifically detects ERK2 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
ERK2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-MAPK1-1D1 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2 | |
Recombinant full-length human ERK2 protein (Uniprot: P28482) | |
0.1 mL | |
Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Protein Kinase, Signal Transduction, Tumor Suppressors | |
5594 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction