Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ERK2 C-Term Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992071
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2071 25 μL
50-199-2070 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2071 Supplier Rockland Immunochemicals Supplier No. 600401GP1S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human ERK2 protein. Cross reactivity is expected to occur with human, mouse and rat based on sequence identity of the peptide immunogen. This antibody does not react with the ERK1 isoform.

ERK2 (also known as MAPK1) is a member of the MAP kinase subfamily of Ser/Thr protein kinases. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.

Specifications

Antigen ERK2 C-Term
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene MAPK1
Gene Accession No. P28482, P63085, P63086
Gene Alias 9030612K14Rik; AA407128; AU018647; C78273; ERK; Erk2; ERK-2; ERT1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase-2; extracellular-signal-regulated kinase 2; MAP kinase 1; MAP kinase 2; MAP kinase isoform p42; Mapk; MAPK 1; MAPK 2; MAPK1; MAPK2; mitogen activated protein kinase 1; Mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; p38; p40; p41; p41mapk; p42mapk; p42-MAPK; PRKM1; PRKM2; protein tyrosine kinase ERK2
Gene Symbols MAPK1
Host Species Rabbit
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116590, 26413, 5594
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.