Learn More
Promega ERK2 Kinase Enzyme System, 10microg
Catalog No. PRV1961 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV1961 10 μg
1 options
Catalog No. PRV1961 Supplier Promega Supplier No. V1961
Recombinant full-length human ERK2 was expressed by E. coli cells using an N-terminal GST tag and activated by MEK1 in vitro. ERK2 is a protein serine/threonine kinase that is a member of the extracellular signal-regulated kinases (ERKs).
  • Easily Screen and Profile ERK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C