Learn More
Promega ERK2 Kinase Enzyme System, 1mg
Catalog No. PRV1962 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV1962 1 mg
1 options
Catalog No. PRV1962 Supplier Promega Supplier No. V1962
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human ERK2 was expressed by E. coli cells using an N-terminal GST tag and activated by MEK1 in vitro. ERK2 is a protein serine/threonine kinase that is a member of the extracellular signal-regulated kinases (ERKs).
  • Easily Screen and Profile ERK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C