Learn More
Specifications
Specifications
| Gene Accession No. | P28482, P28482, P28482, P28482 |
| Preparation Method | This product has been tested by SDS-PAGE and western blot and is suitable for use in Western blot, ELISA, Immunoprecipitation and ChIP. No detection of expected band at ~44 kDa is observed in MAPK1 (ERK2) knockout A549 when compared with unmodified A549 cell lysates by Western blot. |
| Quantity | 100 μg |
| Purity | Multi-step |
| Host Species | Human |
| Content And Storage | Store at -70°C. Avoid freeze/thaw cycles. |
| Product Line | Novus |
| Research Category | Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Tumor Suppressors |
| Target | ERK2 |
| Synonym | EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
