Learn More
Invitrogen™ ERK2 Monoclonal Antibody (4C11)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515304
Description
MA5-15304 targets ERK2 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human, mouse, and Non-human primate samples. The MA5-15304 immunogen is purified recombinant fragment of human ERK2 expressed in E. Coli. MA5-15304 detects ERK2 which has a predicted molecular weight of approximately 41kDa.
MAPK1 (ERK2) is a serine/threonine kinase that acts as an important activator of p90 RSK, MSK, ELK1, and Stat3, and is involved in the MAPK pathway. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. The activation of this kinase requires its phosphorylation by upstream kinases. Upon activation, this kinase translocates to the nucleus of the stimulated cells, where it phosphorylates nuclear targets.
Specifications
| ERK2 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.03% sodium azide | |
| P28482, P63085 | |
| MAPK1 | |
| Purified recombinant fragment of human ERK2 expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG2a |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4C11 | |
| Unconjugated | |
| MAPK1 | |
| 9030612K14Rik; AA407128; AU018647; C78273; ERK; ERK2; ERK-2; ERT1; extracellular signal-regulated kinase 2; extracellular signal-regulated kinase-2; extracellular-signal-regulated kinase 2; MAP kinase 1; MAP kinase 2; MAP kinase isoform p42; Mapk; MAPK 1; MAPK 2; Mapk1; MAPK2; mitogen activated protein kinase 1; mitogen-activated protein kinase 1; Mitogen-activated protein kinase 2; p38; p40; p41; p41mapk; P42MAPK; p42-MAPK; Prkm1; PRKM2; protein tyrosine kinase ERK2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 26413, 5594 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.